Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548464
Description
TAC4 Polyclonal Antibody for Western Blot
Tachykinins are active peptides which excite neurons, evoke behavioral responses, are potent vasodilators and secretagogues, and contract (directly or indirectly) many smooth muscles. Endokinin-A induces thermal hyperalgesia and pain-related behavior such as scratching following intrathecal administration in rats. These effects are suppressed by treatment with endokinin- C. Endokinin-A/B reduces arterial blood pressure and increases sperm motility.
Specifications
TAC4 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q86UU9 | |
TAC4 | |
KLH conjugated synthetic peptide between 85-113 amino acids from the C-terminal region of human TAC4. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TAC4 | |
EK; EKA; EKA/B; EKC; endokinin; Endokinin-A; Endokinin-A/B; Endokinin-C; HK1; HK-1; PPT-C; Preprotachykinin-C; TAC4; tachykinin 4 (hemokinin); tachykinin-4 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
255061 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction