Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TACC1 Monoclonal Antibody (OTI1H2)
GREENER_CHOICE

Catalog No. PIMA526730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526730 Supplier Invitrogen™ Supplier No. MA526730
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TACC1 Monoclonal Antibody for Western Blot, IHC (P)

This locus may represent a breast cancer candidate gene. It is located close to FGFR1 on a region of chromosome 8 that is amplified in some breast cancers. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TACC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TACC1
Gene Accession No. O75410
Gene Alias 4833447E04Rik; AA960202; B230378H13Rik; Ga55; gastric cancer antigen Ga55; KIAA1103; Tacc1; Tacc1a; Taxin-1; transforming acidic coiled-coil containing protein 1; transforming acidic coiled-coil-containing protein 1; transforming, acidic coiled-coil containing protein 1; transforming, acidic coiled-coil containing protein 1a
Gene Symbols TACC1
Host Species Mouse
Immunogen Full length human recombinant protein of TACC1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6867
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.