Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988143
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8143 Supplier Bioss Supplier No. BS7939R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This locus may represent a breast cancer candidate gene. It is located close to FGFR1 on a region of chromosome 8 that is amplified in some breast cancers. Three transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen TACC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TACC1
Gene Accession No. O75410
Gene Alias 4833447E04Rik; AA960202; B230378H13Rik; Ga55; Gastric cancer antigen Ga55; KIAA1103; TACC1; Taxin-1; transforming acidic coiled-coil containing protein 1; Transforming acidic coiled-coil-containing protein 1; transforming, acidic coiled-coil containing protein 1
Gene Symbols TACC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TACC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6867
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.