Learn More
TACC2 Mouse anti-Human, Clone: OTI8A5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804400

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Transforming acidic coiled-coil proteins are a conserved family of centrosome- and microtubule-interacting proteins that are implicated in cancer. This gene encodes a protein that concentrates at centrosomes throughout the cell cycle. This gene lies within a chromosomal region associated with tumorigenesis. Expression of this gene is induced by erythropoietin and is thought to affect the progression of breast tumors. Several transcript variants encoding different isoforms have been found for this gene.Specifications
TACC2 | |
Monoclonal | |
Unconjugated | |
TACC2 | |
AZU-1, ECTACC | |
Mouse | |
Affinity Chromatography | |
RUO | |
10579 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8A5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95359 | |
TACC2 | |
Human recombit protein fragment corresponding to amino acids 727-1026 of human TACC2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.