Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TACC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556617
Description
Immunogen sequence: NLPTHGGQEQ ALGSELQSQL PKGTLSDTPT SSPTDMVWES SLTEESELSA PTRQKLPALG EKRPEGACGD GQSSRVSPPA ADVLKDFSLA GNFS Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 41%.
Transforming acidic coiled-coil proteins are a conserved family of centrosome- and microtubule-interacting proteins that are implicated in cancer. This gene encodes a protein that concentrates at centrosomes throughout the cell cycle. This gene lies within a chromosomal region associated with tumorigenesis. Expression of this gene is induced by erythropoietin and is thought to affect the progression of breast tumors. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
TACC2 | |
Polyclonal | |
Unconjugated | |
TACC2 | |
anti zuai-1; Anti-Zuai-1; AZU-1; ECTACC; mKIAA4180; MNCb-3527; RP11-296H2.1; TACC2; transforming acidic coiled coil 2; transforming acidic coiled-coil containing protein 2; transforming acidic coiled-coil containing protein 2 variant 3; transforming acidic coiled-coil-containing protein 2; transforming, acidic coiled-coil containing protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10579 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95359 | |
TACC2 | |
Recombinant protein corresponding to Human TACC2. Recombinant protein control fragment (Product #RP-94988). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction