Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TACC3 Antibody (OTI6F1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274436B
Description
TACC3 Monoclonal specifically detects TACC3 in Human, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TACC3 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
TACC3 | |
Full length recombinant protein of human TACC3 (NP_006333) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Primate, Monkey | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI6F1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ERIC1ERIC-1, MGC117382, MGC133242, transforming acidic coiled-coil-containing protein 3, transforming, acidic coiled-coil containing protein 3 | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer, Mitotic Regulators, Tumor Suppressors | |
10460.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction