Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TACE Human Recombinant anti-Human, Clone: D1(A12), MilliporeSigma™
Human Recombinant Monoclonal Antibody
Supplier: MilliporeSigma MABT884
Description
Detect TACE/ADAM17 using this human monoclonal Anti-TACE, clone D1(A12) validated for use in Flow Cytometry and Activity/Function Inhibition.
Specifications
TACE | |
Monoclonal | |
Please refer to lot specific datasheet. | |
ADAM17,CSVP,TACE | |
See Specificity section. | |
RUO | |
Primary | |
Cat. No. MABT884 is a recombinant human IgG1 that contains a heavy chain variable (VH) region with affinity towards human TACE noncatalytic Dis-Cys domain and a light chain variable (VL) region that binds human TACE catalytic domain (Tape, C.J., et al. (2011). Proc. Natl. Acad. Sci. U. S. A. 108(14):5578-5583). The VH region of D1(A12) is derived from the scFv named D1 isolated from a phage library generated using pooled lymphocytes from 43 donors (Schofield, D.J., et al. (2007). Genome Biol. 8(11):R254). The D1 VH cDNA was subsequently cloned into a naïve human VL phage-display library for identification and isolation of the hybrid D1(A12) scFv with affinity toward both Dis-Cys domain and the catalytic domain. The D1 VH and A12 VL cDNAs were then cloned into a pBudCE4.1 human IgG1 expression vector for generating the recombinant antibody in HEK293 cells (Tape, C.J., et al. (2011). Proc. Natl. Acad. Sci. USA. 108(14):5578-5583). | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
IgG1 κ |
Inhibition Assays, Flow Cytometry | |
D1(A12) | |
Unconjugated | |
Human | |
100 μg | |
Apoptosis and Cancer | |
NP_003174 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction