Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TACI Polyclonal Antibody
TACI Polyclonal Antibody
Supplier: Thermo Scientific PA580147
Description
TACI Polyclonal Antibody for Western Blot, ELISA

Specifications
TACI | |
Polyclonal | |
Unconjugated | |
TNFRSF13B | |
1200009E08Rik; CD267; CVID; CVID2; IGAD2; NEWGENE_1595998; RYZN; TACI; TNF receptor superfamily member 13B; Tnfrsf13b; TNFRSF14B; transmembrane activator and calcium-modulating and cyclophilin ligand interactor; transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23495 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
O14836 | |
TNFRSF13B | |
E. coli-derived human TACI recombinant protein (Position: M1-S165). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction