Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TACI/TNFRSF13B/CVID Antibody (006), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290025AF647
Description
TACI/TNFRSF13B/CVID Monoclonal specifically detects TACI/TNFRSF13B/CVID in Human samples. It is validated for ELISA.Specifications
TACI/TNFRSF13B/CVID | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
6 | |
ELISA | |
CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TACI/TNFRSF13B/CVID (O14836-2; Ser2-Thr120). | |
0.1 mL | |
Apoptosis, B Cell Development and Differentiation Markers, Signal Transduction | |
23495 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction