Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TACI/TNFRSF13B/CVID Antibody (166010), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1041D
missing translation for 'nonReturnableMessage'
missing translation for 'returnPolicyLabel'
Description
TACI/TNFRSF13B/CVID Monoclonal antibody specifically detects TACI/TNFRSF13B/CVID in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA (Matched Antibody Pair)Specifications
TACI/TNFRSF13B/CVID | |
Monoclonal | |
DyLight 350 | |
CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B | |
Mouse myeloma cell line NS0-derived recombinant mouse TACI/TNFRSF13B, Phe5-Thr129, Accession # Q9ET35 | |
0.5 mL | |
Apoptosis, B Cell Development and Differentiation Markers, Signal Transduction | |
23495 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, ELISA | |
166010 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction