Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TACI/TNFRSF13B/CVID Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TACI/TNFRSF13B/CVID |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TACI/TNFRSF13B/CVID Polyclonal specifically detects TACI/TNFRSF13B/CVID in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TACI/TNFRSF13B/CVID | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
23495 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ACFLKKRGDPCSCQPRSRPRQSPAKSSQDHAMEAGSPVSTSPEPVETCSFCFPECRAPTQESAVTPGTPDPTCAGRWGCHTRTTVLQPCPHIPDSGLGIVCVPAQE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B | |
TNFRSF13B | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title