Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518712
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518712 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518712 Supplier Thermo Scientific Supplier No. PA518712
Add to Cart
Add to Cart

TACR1 Polyclonal Antibody

TACR1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TACR1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Tacr1
Gene Accession No. P25103, P30548
Gene Alias neurokinin 1 receptor; neurokinin receptor 1; neurokinin receptor subtype NK1; neurokinin-1 receptor; Nk1 receptor; NK-1 receptor; Nk1r; NK-1R; NKIR; SPR; substance p receptor; substance-P receptor; TAC1R; tachykinin receptor 1; tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor); TACR1; truncated neurokinin-1 receptor
Gene Symbols Tacr1
Host Species Goat
Immunogen Synthetic peptide sequence (EIPGDSSDRYHEQ) corresponding to the internal amino acids of TACR1 (aa 227-240).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21336, 6869
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.