Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TACR2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550560

Catalog No. PA550560


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total Nk2 / TACR2 protein.

This gene belongs to a family of genes that function as receptors for tachykinins. Receptor affinities are specified by variations in the 5'-end of the sequence. The receptors belonging to this family are characterized by interactions with G proteins and 7 hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin neuropeptide substance K, also referred to as neurokinin A.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TACR2
Polyclonal
Unconjugated
TACR2
7 transmembrane receptor; neurokinin 2 receptor; neurokinin A receptor; NK2; NK-2 receptor; NK2R; NK-2R; NKNAR; seven transmembrane helix receptor; SK receptor; SKR; substance K receptor; substance-K receptor; TAC2R; Tachykinin 2 (substance K) receptor; tachykinin NK-2 receptor; tachykinin receptor 2; TACR2
Rabbit
Antigen affinity chromatography
RUO
6865
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P21452
TACR2
Synthetic peptide corresponding to residues near the N terminal of human tachykinin receptor 2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.