Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TACR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA591934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA591934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA591934 Supplier Invitrogen™ Supplier No. PA591934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLAFRCCPWV TPTKEDKLEL TPTTSLSTRV NRCHTKETLF MAGDTAPSEA TSGEAGRPQD GSGLWFGYGL LAPTKTHVEI; Positive Samples: SGC-7901, A-549; Cellular Location: Cell membrane, Multi-pass membrane protein.

This gene belongs to a family of genes that function as receptors for tachykinins. Receptor affinities are specified by variations in the 5'-end of the sequence. The receptors belonging to this family are characterized by interactions with G proteins and 7 hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin neuropeptide substance K, also referred to as neurokinin A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TACR2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TACR2
Gene Accession No. P21452
Gene Alias 7 transmembrane receptor; neurokinin 2 receptor; neurokinin A receptor; NK2; NK-2 receptor; NK2R; NK-2R; NKNAR; seven transmembrane helix receptor; SK receptor; SKR; substance K receptor; substance-K receptor; TAC2R; Tachykinin 2 (substance K) receptor; tachykinin NK-2 receptor; tachykinin receptor 2; TACR2
Gene Symbols TACR2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 319-398 of human TACR2 (NP_0010482).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6865
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.