Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TACR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111739 Supplier Invitrogen™ Supplier No. PA5111739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STVTMDQGAT KCVVAWPEDS GGKTLLL Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 78%.

This gene belongs to a family of genes that function as receptors for tachykinins. Receptor affinities are specified by variations in the 5'-end of the sequence. The receptors belonging to this family are characterized by interactions with G proteins and 7 hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin neuropeptide substance K, also referred to as neurokinin A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TACR2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TACR2
Gene Accession No. P21452
Gene Alias 7 transmembrane receptor; neurokinin 2 receptor; neurokinin A receptor; NK2; NK-2 receptor; NK2R; NK-2R; NKNAR; seven transmembrane helix receptor; SK receptor; SKR; substance K receptor; substance-K receptor; TAC2R; Tachykinin 2 (substance K) receptor; tachykinin NK-2 receptor; tachykinin receptor 2; TACR2
Gene Symbols TACR2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TACR2. Recombinant protein control fragment (Product #RP-108663).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6865
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.