Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TACR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534249
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534249 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534249 Supplier Invitrogen™ Supplier No. PA534249
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%) Gibbon, Marmoset (94%) Mouse, Rat, Hamster, Panda, Bovine, Dog, Rabbit (88%).

The tachykinins are a family of small peptides that include the neurotransmitters substance P, neurokinin A, and neurokinin B, which can act on three related but distinct seven transmembrane G-proteins coupled receptors, albeit at different concentrations. The NK-3 receptor (NK3R) has greatest affinity for neurokinin B and is highly expressed in the supraoptic and paraventricular nuclei. Following binding of its ligand, NK3R activates a phosphatidylinositol-calcium second messenger system. It is likely these signals lead to the release of vasopressin and oxytocin into the circulation. NK3R may be involved in learning and memory as mice lacking this gene expressed cognitive deficits compared to normal mice. Although it has been suggested that NK3R plays a role in the regulation of vagal afferent relay neurons, it is likely that these receptors are activated by substance P or neurokinin A, as the airway nerves do not express neurokinin B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TACR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Tacr3
Gene Accession No. P29371
Gene Alias G protein-coupled receptor; HH11; neurokinin 3 receptor; Neurokinin B; neurokinin B receptor; neurokinin beta receptor; neuromedin K receptor; Neuromedin K receptor (Neurokinin B/Tachikin 3); neuromedin-K receptor; Nk3 receptor; NK-3 receptor; NK3R; NK-3R; NK3R TAC3R; Nkr; Nmkr; Nmk-r; TAC3R; TAC3RL; tachikin receptor 3; tachykinin receptor 3; Tacr3
Gene Symbols Tacr3
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from C-terminal cytoplasmic domain of human TACR3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6870
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.