Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TACS Nuclease and Buffer
SDP

Catalog No. 480030N Shop All Novus Products
Change view
Click to view available options
Quantity:
15 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
480030N 15 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 480030N Supplier R&D Systems™ Supplier No. 480030N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly purified recombinant endonuclease specific for all forms of DNA

  • TACS-Nuclease has high specific activity, is tolerant to a variety of buffer conditions, and is stable at both 4°C and at -20°C
  • TACS-Nuclease Buffer has been optimized for use with TACS-Nuclease for the generation of a positive control for DNA fragmentation characteristic of apoptosis

Specifications

Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 15 μL
Product Type TACS Nuclease and Buffer
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.