Description
- Use substrate solution at room temperature, add 100μL of solution per well.
- TACS-Sapphire™ is supplied ready to use.
- Protect from light.
- Non-toxic, non-organic peroxidase substrate.
Specifications
Specifications
| Quantity | 10 mL |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |