Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tacstd1 Mouse anti-Rat, Clone: Epcam/1158, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14196
Description
Mouse monoclonal antibody raised against full length recombinant rat Tacstd1.
- High specificity
- High reactivity
Specifications
Tacstd1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (5-10 uL/million cells in 0.1 mL) Immunofluorescence (1:50-1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) Western Blot (1:100-1:200) The optimal working dilution should be determined by | |
O55159 | |
Tacstd1 | |
Recombinant protein corresponding to full length rat Tacstd1. | |
RUO | |
171577 | |
Store at 4°C. | |
IgG1 |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Epcam/1158 | |
Mouse monoclonal antibody raised against full length recombinant rat Tacstd1. | |
In bioreactor concentrate (0.05% sodium azide). | |
Egp314 | |
Mouse | |
500 μL | |
Primary | |
Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction