Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tacstd1 Mouse anti-Rat, Clone: Epcam/1159, Abnova™

Catalog No. 89034298 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-298 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-298 Supplier Abnova Corporation Supplier No. MAB14197
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant rat Tacstd1.

  • High specificity
  • High reactivity

Specifications

Antigen Tacstd1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone Epcam/1159
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant rat Tacstd1.
Dilution Flow Cytometry (5-10 uL/million cells in 0.1 mL) Immunofluorescence (1:50-1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) Western Blot (1:100-1:200) The optimal working dilution should be determined by
Formulation In bioreactor concentrate (0.05% sodium azide).
Gene Accession No. O55159
Gene Alias Egp314
Gene Symbols Tacstd1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length rat Tacstd1.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171577
Target Species Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.