Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACSTD2 Mouse anti-Human, Unconjugated, Clone: TACSTD2/2151, Abnova™

Catalog No. 89337777 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-777 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-777 Supplier Abnova Corporation Supplier No. MAB21286
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

TACSTD2 Monoclonal antibody specifically detects TACSTD2 in Human samples. It is validated for WB, IHC-P.

Specifications

Antigen TACSTD2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TACSTD2/2151
Conjugate Unconjugated
Description tumor-associated calcium signal transducer 2
Gene TACSTD2
Gene Alias EGP-1, GA733, GA733-1, M1S1, TROP2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 31-274 of human TACSTD2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4070
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.