Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TACTILE (CD96) Mouse anti-Human, BB515, Clone: NK92.39.rMAb, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568847
Description
TACTILE (CD96) Monoclonal antibody specifically detects TACTILE (CD96) in Human samples. It is validated for Flow cytometry
Specifications
TACTILE (CD96) | |
Monoclonal | |
Brilliant Blue 515 (BB515) | |
hCD96; T cell-activated increased late expression protein | |
Human NK92 Cell Line | |
100 Tests | |
Primary | |
Human | |
Liquid |
Flow Cytometry | |
NK92.39.rMAb | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
10225 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction