Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TADA1L Antibody (PCRP-TADA1-1C9), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314264MFV610
Description
TADA1L Monoclonal antibody specifically detects TADA1L in Human samples. It is validated for Flow CytometrySpecifications
TADA1L | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
117143 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry | |
PCRP-TADA1-1C9 | |
Flow Cytometry | |
ADA1, hADA1, HFI1, SPT3-associated factor 42, SPT3-associated factor 42 (STAF42), STAF42transcriptional adaptor 1 (HFI1 homolog, yeast)-like, TADA1LRP1-9E21.4, transcriptional adapter 1, Transcriptional adapter 1-like protein, transcriptional adaptor 1, transcriptional adaptor 1 (HFI1 homolog, yeast) | |
Recombinant full-length human TADA1Lprotein (Uniprot: Q96BN2) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction