Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TADA1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569843 Supplier Invitrogen™ Supplier No. PA569843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 75%.

Probably involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TADA1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TADA1
Gene Accession No. Q96BN2
Gene Alias 2900026B15Rik; ADA1; D1Ertd251e; hADA1; HF1L; HFI1; RGD1306774; SPT3-associated factor 42; SPT3-associated factor 42 (STAF42); STAF42; TADA1; tada1l; transcriptional adapter 1; Transcriptional adapter 1-like protein; transcriptional adaptor 1; transcriptional adaptor 1 (HFI1 homolog, yeast); transcriptional adaptor 1 (HFI1 homolog, yeast) like; transcriptional adaptor 1- like; zgc:85851
Gene Symbols TADA1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human TADA1L.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117143
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.