Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TADA2B Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TADA2B |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1534
![]() ![]() |
Proteintech Group Inc
173671AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Specifications
TADA2B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q86TJ2 | |
93624 | |
TADA2B Fusion Protein Ag10783 | |
Primary | |
-20°C | |
TADA2B |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ADA2 beta, ADA2 like protein beta, ADA2(beta), ADA2B, TADA2B, Transcriptional adapter 2 beta | |
TADA2B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title