Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TADA2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104485 Supplier Invitrogen™ Supplier No. PA5104485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ADA2L.

TADA2L (transcriptional adapter 2-like), also known as TADA2A (transcriptional adapter 2-alpha) or ADA2-like protein, is a 443 amino acid nuclear protein that exists as 2 alternatively spliced isoforms. While most abundantly expressed in testis, TADA2L is present in all tissues. TADA2L contains one SANT domain and one SWIRM domain, and interacts with GCN5 and GR (NR3C1). Its ability to bind double-stranded DNA allows TADA2L to play a role in chromatin remodeling. Although it makes up part of the PCAF complex, TADA2L is also a component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4. The gene that encodes TADA2L contains 71,408 bases and maps to human chromosome 17q12. Chromosome 7 houses over 1,000 genes, comprises nearly 5% of the human genome and has been linked to osteogenesis imperfecta, Pendred syndrome, lissencephaly, citrullinemia and Shwachman-Diamond syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TADA2L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TADA2A
Gene Accession No. O75478, Q6AYE3, Q8CHV6
Gene Alias ADA2; ADA2A; ADA2-like protein; AV319371; D030022J10Rik; hADA2; KL04P; TADA2A; TADA2L; transcriptional adapter 2-alpha; Transcriptional adapter 2-like; transcriptional adaptor 2 alpha; transcriptional adaptor 2A
Gene Symbols TADA2A
Host Species Rabbit
Immunogen A synthesized peptide derived from human TADA2A(Accession O75478), corresponding to amino acid residues R218-K268.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217031, 360581, 6871
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.