Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tadalafil
GREENER_CHOICESDP

Catalog No. 631110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-111-0 10 mg
63-115-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-111-0 Supplier Tocris Bioscience™ Supplier No. 6311/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and highly selective PDE5 inhibitor; orally bioavailable

Potent and highly selective PDE5 inhibitor (IC50 = 5 nM). Exhibits > 2000-fold selectivity for PDE5 over PDEs 1,2,3 and 4 and 1000-fold selectivity over PDE6. Reduces blood pressure in hypertensive rats. Also delays tumor growth in a mouse model via inhibition of MDSC-mediated immunosuppression. Orally bioavailable.

Chemical Identifiers

CAS 171596-29-5
Molecular Formula C22H19N3O4

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 389.4
Physical Form Solid
Purity >99%
Molecular Weight (g/mol) 389.411
Chemical Name or Material (6R,12aR)-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-methylpyrazino[1',2':1,6]pyrido[3,4-b]
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.