Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAE 50X BUFFER, MP Biomedicals™

Catalog No. MP1TAE50X01 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1TAE50X01 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1TAE50X01 Supplier MP Biomedicals, Inc Supplier No. 11TAE50X01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TAE 50X BUFFER

  • TAE running buffer is the most commonly used buffer for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis.
  • Buffer for Electrophoresis.
  • Prepare electrophoresis buffers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00236357
Synonym Tris-Borate-EDTA Buffer

Specifications

Color Clear Colorless
CAS 7732-18-5
For Use With (Application) Agarose Gel Electrophoresis, Polyacrylamide Gel Electrophoresis
pH 8.2 to 8.4
Physical Form Solution
Quantity 1 L
Solution Type Buffer
Formulation Tris Base: 2M; Acetic Acid : 1M; EDTA : 0.05 M Acetic acid: 1M EDTA: 0.05M
Grade Molecular Biology
storageNote1 Store at 2-8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.