Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAE, 50X, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2 L
4 L
Description
General Description
- TAE (50X), TRIS + acetate + EDTA allows nucleic acid separation
Application
- This buffer has been used to prepare agarose and polyacrylamide gels for electrophoresis of nucleic acids
- It is used as a running and gel preparation buffer, and it is also used for non-denaturing RNA agarose gel electrophoresis

Specifications
Specifications
Chemical Name or Material | Buffer |
Concentration | 50X |
Physical Form | Liquid |
Quantity | 1 L |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction