Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
5 L
Packaging:
Nalgene bottle
Upright carton
Specifications
Specifications
Chemical Name or Material | TAE Buffer |
Concentration | 10X |
Physical Form | Solution |
Quantity | 1 L |
Packaging | Nalgene bottle |
Grade | Molecular Biology, Ultrapure |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction