Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119850
This item is not returnable.
View return policy
Description
Positive Control: Rat kindey tissue, mouse colon tissue. Subcellular Location: Nucleus.
3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.
Specifications
TAF1 | |
Polyclonal | |
Unconjugated | |
TAF1 | |
AU015687; B430306D02Rik; BA2R; Ccg1; Ccg-1; CCGS; Cell cycle gene 1 protein; cell cycle, G1 phase defect; complementation of cell cycle block, G1-to-S; DYT3; DYT3/TAF1; KAT4; MRXS33; NSCL2; N-TAF1; OF; P250; RGD1562050; TAF(II)250; TAF1; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, neuron specific isoform; Taf2a; TAFII250; TAFII-250; TATA box binding protein associated factor 1; TATA-box binding protein associated factor 1; TBP-associated factor 250 kDa; transcription factor TFIID p250 polypeptide; transcription initiation factor TFIID 250 kDa subunit; Transcription initiation factor TFIID subunit 1; XDP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
270627, 317256, 6872 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P21675, Q80UV9 | |
TAF1 | |
Recombinant protein within human aa 1500-1700. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction