Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF10 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992126
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2126 25 μL
50-199-2125 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2126 Supplier Rockland Immunochemicals Supplier No. 600401GW8S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity-purified antibody is directed against human TAF10 protein. A BLAST analysis was used to suggest cross reactivity with TAF10 protein from human and mouse (100% homology). Reactivity against homologues from other sources is not known.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the small subunits of TFIID that is associated with a subset of TFIID complexes. Studies with human and mammalian cells have shown that this subunit is required for transcriptional activation by the estrogen receptor, for progression through the cell cycle, and may also be required for certain cellular differentiation programs.

Specifications

Antigen TAF10
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene TAF10
Gene Accession No. Q12962
Gene Alias 30kDa; AU041226; BOS_14631; im:6899389; LOC100356523; mTAFII30; si:dkey-72l14.8; STAF28; TAF(II)30; Taf10; TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 30 kDa; TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 30kDa; TAF2A; TAF2H; TAFII30; TAFII-30; TATA box binding protein (TBP)-associated factor, RNA polymerase II, H, 30 kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, H, 30kD; TATA box binding protein associated factor 10; TATA-box binding protein associated factor 10; TBP-related factor 10; transcription initiation factor TFIID 30 kD subunit; Transcription initiation factor TFIID 30 kDa subunit; Transcription initiation factor TFIID subunit 10
Gene Symbols TAF10
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6881
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.