Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103318
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103318 Supplier Invitrogen™ Supplier No. PA5103318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAF10.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the small subunits of TFIID that is associated with a subset of TFIID complexes. Studies with human and mammalian cells have shown that this subunit is required for transcriptional activation by the estrogen receptor, for progression through the cell cycle, and may also be required for certain cellular differentiation programs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAF10
Gene Accession No. Q12962, Q8K0H5
Gene Alias 30kDa; AU041226; im:6899389; mTAFII30; si:dkey-72l14.8; STAF28; TAF(II)30; TAF10; TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 30 kDa; TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 30kDa; TAF2A; Taf2h; TAFII30; TAFII-30; TATA box binding protein (TBP)-associated factor, RNA polymerase II, H, 30 kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, H, 30kD; TATA box binding protein associated factor 10; TATA-box binding protein associated factor 10; TBP-related factor 10; transcription initiation factor TFIID 30 kD subunit; transcription initiation factor TFIID 30 kDa subunit; Transcription initiation factor TFIID subunit 10
Gene Symbols TAF10
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAF10(Accession Q12962), corresponding to amino acid residues P14-L64.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24075, 6881
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.