Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TAF10/TAFII30, Mouse, Unlabeled, Clone: 23TA-1H8,

Catalog No. MABE1078MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE1078MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1078MI Supplier MilliporeSigma™ Supplier No. MABE1078
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transcription initiation factor TFIID subunit 10 (UniProt Q12962; also known as STAF28, TAF(II)30, TAFII-30, TAFII30, Transcription initiation factor TFIID 30kDa subunit) is encoded by the TAF10 (also known as TAF2A, TAF2H, TAFII30) gene (Gene ID 6881) in human.Transcription initiation factor TFIID subunit 10 (UniProt Q12962; also known as STAF28, TAF(II)30, TAFII-30, TAFII30, Transcription initiation factor TFIID 30kDa subunit) is encoded by the TAF10 (also known as TAF2A, TAF2H, TAFII30) gene (Gene ID 6881) in human. TAF10 (TAFII30) is a subunit of both the general transcription factor (GTF) TFIID and the coactivator SAGA (Spt-Ada-Gcn5-acetyltransferase) histone acetyltransferase (HAT) complexes involved in the initiation of RNA polymerase II (Pol II) transcription in eukaryotes. TFIID complex is composed of the TATA-binding protein (TBP) and thirteen TBP-associated factors (TAFs) that allow its recognition of not only TATA box-containing, but also TATA-less promoters for site-specific assembly of preinitiation complex (PIC). TAF10-knockout studies in mice indicate that TAF10 plays an important role during embryonic development and homeostasis in a tissue-dependent manner. TAF10 ablation is reported to result in severe anemia and lethality at embryonic stage E13.5 due to impaired erythropoiesis. In addition, TAF10 is shown to not only interact with the transcription factor GATA1, but also regulate its transcription as evidenced by its occupancy at GATA1 genomic locus in human fetal erythroid cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF10/TAFII30
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 23TA-1H8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1κ ascites with 0.05% Sodium Azide.
Gene TAF10, TAF2A, TAF2H, TAFII30
Gene Accession No. Q12962
Host Species Mouse
Immunogen Ovalbumin-conjugated linear peptide corresponding to the N-terminal sequence of human TAF10/TAFII30.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_006275
Test Specificity Clone 23TA-1H8 (a.k.a. 1H8, 23TA 1H8, 23TA1H8) recognizes an N-terminal epitope present in human (UniProt Q12962), but not mouse (UniProt Q8K0H5), TAF10 (TAFII30) sequence.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.