Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565044 Supplier Invitrogen™ Supplier No. PA565044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPSDKGGETG ESDETAAVPG DPGATDTDGI PEETDGDADV DLKEAAAEEG ELESQDVSDL TTVEREDSSL LNPAAKKLKI DTKEKKEKKQ KVDEDEIQKM QILVSSFSEE QLNRYEMY.

TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. TAF11 is a small subunit of TFIID that is present in all TFIID complexes and interacts with TBP. This subunit also interacts with another small subunit, TAF13, to form a heterodimer with a structure similar to the histone core structure. Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAF11
Gene Accession No. Q15544
Gene Alias 1110038O14Rik; 28kDa; AI115401; AL024207; AW552398; hypothetical protein LOC507047; MGC:15243; PRO2134; PRO214; TAF(II)28; Taf11; TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 28kDa; TAF2I; TAFII28; TAFII-28; TATA box binding protein (TBP)-associated factor 11; TATA box binding protein (TBP)-associated factor, RNA polymerase II, I, 28kD; TATA box binding protein associated factor 11; TATA-box binding protein associated factor 11; TBP-associated factor 11; TFIID subunit p30-beta; transcription initiation factor TFIID 28 kD subunit; transcription initiation factor TFIID 28 kDa subunit; Transcription initiation factor TFIID subunit 11
Gene Symbols TAF11
Host Species Rabbit
Immunogen Recombinant Human TAF11. Recombinant protein control fragment (Product #RP-104596).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6882
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.