Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986362
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6362 Supplier Bioss Supplier No. BS5142R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAF12 is involved in the control of transcription by RNA polymerase II involves the basal transcription machinery which is a collection of proteins. These proteins with RNA polymerase II, assemble into complexes which are modulated by transactivator proteins that bind to cis-regulatory elements located adjacent to the transcription start site. Some modulators interact directly with the basal complex, whereas others may act as bridging proteins linking transactivators to the basal transcription factors. Some of these associated factors are weakly attached while others are tightly associated with TBP in the TFIID complex. Among the latter are the TAF proteins. Different TAFs are predicted to mediate the function of distinct transcriptional activators for a variety of gene promoters and RNA polymerases. TAF12 interacts directly with TBP as well as with TAF2I.

Specifications

Antigen TAF12
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TAF12
Gene Accession No. Q16514
Gene Alias 20kDa; 2810422D08Rik; AW557038; BOS_2382; LOC100352341; TAF12; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20 kD; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20 kDa; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20kDa; TAF15; TAF2J; TAFII20; TAFII-20; TAFII20/TAFII15; TAFII-20/TAFII-15; TATA box binding protein (TBP)-associated factor, RNA polymerase II, J, 20kD; TATA box binding protein associated factor 12; TATA-box binding protein associated factor 12; Transcription initiation factor TFIID 20 kDa subunits; Transcription initiation factor TFIID 20/15 kDa subunits; Transcription initiation factor TFIID subunit 12
Gene Symbols TAF12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TAF12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6883
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.