Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560728 Supplier Invitrogen™ Supplier No. PA560728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDPTFEEENE EIGGGAEGGQ GKRKRLFSKE LRCMMYGFGD DQNPYTESVD ILEDLVIEFI TEMTHKAMSI GRQGRVQVED IVFLIRKDPR KFARVKDLLT MNEELKRA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a small subunit associated with a subset of TFIID complexes. This subunit interacts with TBP and with two other small subunits of TFIID, TAF10 and TAF11. There is a pseudogene located on chromosome 6. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF13
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAF13
Gene Accession No. Q15543
Gene Alias 1810004N01Rik; 2010309N11Rik; AI847295; MGC22425; TAF(II)18; TAF13; TAF13 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF13 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 18kDa; TAF13 RNA polymerase II, TATA box-binding protein-associated factor; Taf2k; TAFII18; TAFII-18; TATA box binding protein (TBP)-associated factor, RNA polymerase II, K, 18kD; TATA box binding protein associated factor 13; TATA-box binding protein associated factor 13; TBP-associated factor, RNA polymerase II, K, 18kD; transcription initiation factor TFIID 18 kD subunit; transcription initiation factor TFIID 18 kDa subunit; Transcription initiation factor TFIID subunit 13
Gene Symbols TAF13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAF13. Recombinant protein control fragment (Product #RP-89444).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6884
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.