Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF15 Antibody (4D71), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192685AF532
Description
TAF15 Monoclonal antibody specifically detects TAF15 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
TAF15 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
4D71 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
68kDa, hTAFII68, Npl3, RBP56TAFII68, RNA-binding protein 56, TAF(II)68, TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2NRBP56/CSMF fusion, TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD(RNA-binding protein 56), TATA box-binding protein-associated factor 2N (RNA-binding protein 56), TATA-binding protein-associated factor 2N, TBP-associated factor 15,68 kDa TATA-binding protein-associated factor | |
Full length recombinant human TAF15 expressed in and purified from E. coli. [UniProt# Q92804] | |
0.1 mL | |
DNA Repair, Neuroscience | |
8148 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction