Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF15 Antibody (SR2205), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322496100UL
Description
TAF15 Monoclonal antibody specifically detects TAF15 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
TAF15 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR2205 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
68kDa, hTAFII68, Npl3, RBP56TAFII68, RNA-binding protein 56, TAF(II)68, TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2NRBP56/CSMF fusion, TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD(RNA-binding protein 56), TATA box-binding protein-associated factor 2N (RNA-binding protein 56), TATA-binding protein-associated factor 2N, TBP-associated factor 15,68 kDa TATA-binding protein-associated factor | |
A synthesized peptide derived from human TAF15 (Uniprot #: Q92804) | |
100 μg | |
DNA Repair, Neuroscience | |
8148 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction