Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF15 Monoclonal Antibody (4D71)
GREENER_CHOICE

Catalog No. PIMA547431
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547431 100 μL
PIMA547332 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA547431 Supplier Invitrogen™ Supplier No. MA547431
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAF15 Monoclonal Antibody for Western Blot, ICC/IF

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF15
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4D71
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene TAF15
Gene Accession No. Q92804
Gene Alias 2610111C21Rik; 68 kDa TATA-binding protein-associated factor; 68kDa; hTAFII68; LOC100713096; Npl3; RBP56; RBP56/CSMF fusion; RNA-binding protein 56; TAF(II)68; Taf15; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68 kDa; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68kDa; TAF2N; TAFII68; TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD (RNA binding protein 56); TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD (RNA-binding protein 56); TATA box binding protein associated factor 15; TATA box-binding protein-associated factor 2N (RNA-binding protein 56); TATA-bin; TATA-binding protein-associated factor 2N; TATA-box binding protein associated factor 15; TBP-associated factor 15
Gene Symbols TAF15
Host Species Mouse
Immunogen Full length recombinant human TAF15 expressed in and purified from E. coli.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287571, 70439, 8148
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.