Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF172 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567118
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567118 Supplier Invitrogen™ Supplier No. PA567118
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QFAARYGKPIL ASRDARSSSR EQEAGVLAMD ALHRQVLPFL LRRMKEDVLQ DLPPKIIQDY YCTLSPLQVQ LYEDFAKSRA KCDVDETVSS ATLS Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

Initiation of transcription by RNA polymerase II requires the assistance of TATA box-binding protein (TBP) and TBP-associated factors, or TAFs (e.g., TAF2B), in 2 distinct complexes, TFIID and B-TFIID. The TFIID complex is composed of TBP and more than 8 TAFs. However, the majority of TBP is present in the B-TFIID complex, which is composed of TBP and TAFII170, also called TAF172, and has DNA-dependent ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF172
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BTAF1
Gene Accession No. O14981
Gene Alias AI414500; AI447930; ATP-dependent helicase BTAF1; BTAF1; BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, (Mot1 homolog, S. cerevisiae); BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa; BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1 homolog, S. cerevisiae); B-TFIID TATA-box binding protein associated factor 1; B-TFIID transcription factor-associated 170 kDa subunit; E430027O22Rik; MOT1; Mot1 homolog; RGD1564130; TAF(II)170; TAF170; TAF172; TAF-172; TAFII170; TATA-binding protein-associated factor 172; TBP-associated factor 170; TBP-associated factor 172
Gene Symbols BTAF1
Host Species Rabbit
Immunogen Recombinant Human TAF172. Recombinant protein control fragment (Product #RP-107762).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9044
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.