Learn More
Invitrogen™ TAF172 Polyclonal Antibody

Description
Positive Samples: HepG2, HeLa, MCF7, Rat testis; Cellular Location: Nucleus.
Specifications
Specifications
| Antigen | TAF172 |
| Applications | ChIP Assay, ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.57 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | BTAF1 |
| Gene Accession No. | O14981 |
| Gene Alias | AI414500; AI447930; ATP-dependent helicase BTAF1; BTAF1; BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, (Mot1 homolog, S. cerevisiae); BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa; BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1 homolog, S. cerevisiae); B-TFIID TATA-box binding protein associated factor 1; B-TFIID transcription factor-associated 170 kDa subunit; E430027O22Rik; MOT1; Mot1 homolog; RGD1564130; TAF(II)170; TAF170; TAF172; TAF-172; TAFII170; TATA-binding protein-associated factor 172; TBP-associated factor 170; TBP-associated factor 172 |
| Gene Symbols | BTAF1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.