Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546849 Supplier Invitrogen™ Supplier No. PA546849
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CEKAFVAGLL LGKGCRYFRY ILKQDHQILG KKIKRMKRSV KKYSIVNPRL Sequence homology: Cow: 78%; Dog: 85%; Horse: 85%; Human: 100%; Mouse: 85%; Pig: 85%; Rabbit: 85%; Rat: 85%.

Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, also known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the smallest SL1-specific TAF. Two transcripts encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF1A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TAF1A
Gene Accession No. Q15573
Gene Alias MGC:17061; mTAFI48; RAFI48; RNA polymerase I-specific TBP-associated factor 48 kDa; similar to TBP-associated factor 1A isoform 1; SL1; SL1, 48kD subunit; Taf1a; TAFI48; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, A; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kDa; TATA box binding protein associated factor 1A; TATA box binding protein-associated factor TAF1A; TATA box-binding protein-associated factor 1A; TATA box-binding protein-associated factor RNA polymerase I subunit A; TATA-box binding protein associated factor, RNA polymerase I subunit A; TATA-box binding protein associated factor, RNA polymerase I, A; TBP-associated factor 1A; Transcription factor SL1; transcription initiation factor SL1/TIF-IB subunit A
Gene Symbols TAF1A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TAF1A (aa 401-450).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9015
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.