Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF1C Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP212779FR
Description
TAF1C Polyclonal antibody specifically detects TAF1C in Human samples. It is validated for Western BlotSpecifications
TAF1C | |
Polyclonal | |
Western Blot | |
MGC:39976, RNA polymerase I-specific TBP-associated factor 110 kDa, SL1, 110kD subunit, SL1TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD, TAFI110TBP-associated factor 1C, TAFI95, TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa, TATA box-binding protein-associated factor 1C, TATA box-binding protein-associated factor RNA polymerase I subunit C, transcription factor SL1, Transcription initiation factor SL1/TIF-IB subunit C | |
The immunogen this antibody was made to, maps to a region between residue 819 and 869 of human TATA Box Binding Protein (TBP)-Associated Factor, RNA Polymerase I, C, 110kDa using the numbering given in entry NP_005670.3 (GeneID 9013). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9013 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction