Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAF1C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577015
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. TAF1C is the largest SL1-specific TAF. Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the largest SL1-specific TAF. Two transcripts encoding different isoforms have been identified.
Specifications
TAF1C | |
Polyclonal | |
Unconjugated | |
TAF1C | |
MGC:39976; mTAFI95; RNA polymerase I-specific TBP-associated factor 110 kDa; RNA polymerase I-specific TBP-associated factor 95 kDa; SL1; SL1, 110kD subunit; TAF1C; TAFI110; TAFI95; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, C; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, C; transcription factor TAFI95; TATA box binding protein associated factor 1C; TATA box-binding protein-associated factor 1C; TATA box-binding protein-associated factor RNA polymerase I subunit C; TATA box-binding protein-associated factor, RNA polymerase I, subunit C; TATA-box binding protein associated factor, RNA polymerase I subunit C; TATA-box binding protein associated factor, RNA polymerase I, C; TBP-associated factor 1C; TBP-associated factor RNA polymerase I 95 kDa; transcription factor SL1; transcription factor TAFI95; transcription initiation factor SL1/TIF-IB subunit C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21341, 361420, 9013 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15572, Q6P773, Q6PDZ2 | |
TAF1C | |
Recombinant full length Human TAF1C. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction