Learn More
Description
Specifications
Specifications
| Antigen | TAF1D |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q9H5J8 |
| Gene Alias | JOSD3RNA polymerase I-specific TBP-associated factor 41 kDa, Josephin domain containing 3, MGC5306, RAFI41, TAF(I)41, TAFI41, TATA box binding protein (TBP)-associated factor, RNA polymerase I, D, 41kDa, TATA box-binding protein-associated factor 1D, TATA box-binding protein-associated factor RNA polymerase I subunit D, TBP-associated factor 1D, Transcription initiation factor SL1/TIF-IB subunit D |
| Gene Symbols | TAF1D |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
