Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF1D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140055 Supplier Invitrogen™ Supplier No. PA5140055
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SLDHVTSDAV ELANRSDNSS DSSLFKTQCI PYSPKGEKRN PIRKFVRTPE SVHASDSSSD SSFEPIPLTI KAIFERFKNR KKRYKKKKKR RYQPTGRPRG RPEGRR Highest antigen sequence indentity to the following orthologs: Mouse - 58%, Rat - 59%.

TAF1D is a member of the SL1 complex, which includes TBP (MIM 600075) and TAF1A (MIM 604903), TAF1B (MIM 604904), and TAF1C (MIM 604905), and plays a role in RNA polymerase I transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF1D
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAF1D
Gene Accession No. Q9H5J8
Gene Alias 2810003M17Rik; 4930553M18Rik; 5930426I11Rik; JOSD3; Josephin domain containing 3; RAFI41; RNA polymerase I-specific TBP-associated factor 41 kDa; TAF(I)41; TAF1D; TAFI41; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, D; TATA box binding protein (TBP)-associated factor, RNA polymerase I, D, 41kDa; TATA box binding protein associated factor 1D; TATA box binding protein-associated factor, RNA polymerase I, D; TATA box-binding protein-associated factor 1D; TATA box-binding protein-associated factor RNA polymerase I subunit D; TATA-box binding protein associated factor, RNA polymerase I subunit D; TATA-box binding protein associated factor, RNA polymerase I, D; TBP-associated factor 1D; Transcription initiation factor SL1/TIF-IB subunit D
Gene Symbols TAF1D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAF1D. Recombinant protein control fragment (Product #RP-108795).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79101
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.