Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAF1D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563212
Description
Immunogen sequence: ESLKQMNVGE DLENEDFDSR RYKFLDDDGS ISPIEESTAE DEDATHLEDN ECDIKLAGDS FIVSSEFPVR LSVYLEEEDI T Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 57%.
TAF1D is a member of the SL1 complex, which includes TBP (MIM 600075) and TAF1A (MIM 604903), TAF1B (MIM 604904), and TAF1C (MIM 604905), and plays a role in RNA polymerase I transcription.
Specifications
TAF1D | |
Polyclonal | |
Unconjugated | |
TAF1D | |
2810003M17Rik; 4930553M18Rik; 5930426I11Rik; JOSD3; Josephin domain containing 3; RAFI41; RNA polymerase I-specific TBP-associated factor 41 kDa; TAF(I)41; TAF1D; TAFI41; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, D; TATA box binding protein (TBP)-associated factor, RNA polymerase I, D, 41kDa; TATA box binding protein associated factor 1D; TATA box binding protein-associated factor, RNA polymerase I, D; TATA box-binding protein-associated factor 1D; TATA box-binding protein-associated factor RNA polymerase I subunit D; TATA-box binding protein associated factor, RNA polymerase I subunit D; TATA-box binding protein associated factor, RNA polymerase I, D; TBP-associated factor 1D; Transcription initiation factor SL1/TIF-IB subunit D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79101 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H5J8 | |
TAF1D | |
Recombinant protein corresponding to Human TAF1D. Recombinant protein control fragment (Product #RP-101444). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction