Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF1D Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA525509

Catalog No. PIPA525509


Only null left
Add to Cart

Description

Description

TAF1D Polyclonal Antibody for Western Blot, IHC (P), Flow

TAF1D is a member of the SL1 complex, which includes TBP (MIM 600075) and TAF1A (MIM 604903), TAF1B (MIM 604904), and TAF1C (MIM 604905), and plays a role in RNA polymerase I transcription.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TAF1D
Polyclonal
Unconjugated
TAF1D
2810003M17Rik; 4930553M18Rik; 5930426I11Rik; JOSD3; Josephin domain containing 3; RAFI41; RNA polymerase I-specific TBP-associated factor 41 kDa; TAF(I)41; TAF1D; TAFI41; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, D; TATA box binding protein (TBP)-associated factor, RNA polymerase I, D, 41kDa; TATA box binding protein associated factor 1D; TATA box binding protein-associated factor, RNA polymerase I, D; TATA box-binding protein-associated factor 1D; TATA box-binding protein-associated factor RNA polymerase I subunit D; TATA-box binding protein associated factor, RNA polymerase I subunit D; TATA-box binding protein associated factor, RNA polymerase I, D; TBP-associated factor 1D; Transcription initiation factor SL1/TIF-IB subunit D
Rabbit
Antigen affinity chromatography, Protein A
RUO
75316, 79101
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9D4V4, Q9H5J8
TAF1D
KLH conjugated synthetic peptide between 62-90 amino acids from the N-terminal region of human TAF1D.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.