Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TAF1L |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TAF1L Polyclonal specifically detects TAF1L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TAF1L | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q8IZX4 | |
138474 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PKQPFMLQHASGEHKDGHGK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
210kDa, 210kDa-like, EC 2.7.11, EC 2.7.11.1, MGC134910, TAF(II)210, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF1-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2A2, TBP-associated factor 1-like, TBP-associated factor 210 kDa, TBP-associated factor RNA polymerase 1-like, Transcription initiation factor TFIID 210 kDa subunit, transcription initiation factor TFIID subunit 1-like | |
TAF1L | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title